Assay Details
| Gene Symbol: AT5G03850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120466.3 | NP_196005.1 | 2 | 241 | 71 | |
| Gene Symbol: AT3G10090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY128400.1 | - | 2 | 250 | 71 | |
| BT000086.1 | - | 1 | 161 | 71 | ||
More Information
Set Membership: |
|

AT5G03850
Genomic Map