Assay Details
| Gene Symbol: ICR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101574.3 | NP_564015.1 | 3 | 970 | 107 | |
| NM_202119.2 | NP_973848.1 | 4 | 932 | 107 | ||
| GenBank mRNA | AY085543.1 | - | 3 | 970 | 107 | |
| BT024727.1 | - | 1 | 675 | 107 | ||
More Information
Set Membership: |
|

ICR1
Genomic Map