Assay Details
| Gene Symbol: PIP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336641.1 | NP_001323849.1 | 4 | 1076 | 156 | |
| NM_129273.5 | NP_181254.1 | 4 | 970 | 156 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIP2B| Gene Symbol: PIP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336641.1 | NP_001323849.1 | 4 | 1076 | 156 | |
| NM_129273.5 | NP_181254.1 | 4 | 970 | 156 | ||
Set Membership: |
|