Assay Details
Gene Symbol: KCS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105524.3 | NP_177020.1 | 2 | 1866 | 148 | |
NM_179530.1 | NP_849861.1 | 1 | 2008 | 148 | ||
GenBank mRNA | AF129511.1 | - | 2 | 1633 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
