Assay Details
| Gene Symbol: AOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113476.4 | NP_566776.1 | 2 | 1092 | 128 | |
| GenBank mRNA | AF380630.1 | - | 2 | 883 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AOC2| Gene Symbol: AOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113476.4 | NP_566776.1 | 2 | 1092 | 128 | |
| GenBank mRNA | AF380630.1 | - | 2 | 883 | 128 | |
Set Membership: |
|