Assay Details
| Gene Symbol: TUB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125665.4 | NP_568960.1 | 3 | 1828 | 115 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length greater than or equal to 101  3' Most  Detect Genomic DNA  Recommended Assay  Arabidopsis thaliana  Both primers and probe map within a single exon | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 TUB3
TUB3 Genomic Map
Genomic Map Back To Top
 Back To Top