Assay Details
Gene Symbol: ARFA1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334452.1 | NP_001323062.1 | 7 | 1161 | 135 | |
NM_001334453.1 | NP_001323063.1 | 7 | 1150 | 135 | ||
NM_001334454.1 | NP_001323060.1 | 7 | 1134 | 135 | ||
NM_001334455.1 | NP_001323061.1 | 7 | 1037 | 135 | ||
NM_105717.3 | NP_177206.1 | 7 | 702 | 135 | ||
NM_180644.3 | NP_850975.1 | 7 | 728 | 135 | ||
NM_202391.2 | NP_974120.1 | 8 | 806 | 135 |
Gene Symbol: ARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF360164.1 | - | 7 | 676 | 135 | |
AY087235.1 | - | 7 | 725 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
