Assay Details
| Gene Symbol: RPS15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331728.1 | NP_001318947.1 | 5 | 636 | 135 | |
| NM_100651.5 | NP_172256.1 | 4 | 572 | 135 | ||
| NM_202054.3 | NP_973783.1 | 4 | 500 | 135 | ||
| Gene Symbol: AT5G59850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF001412.1 | - | 4 | 454 | 135 | |
| AF386922.1 | - | 4 | 461 | 135 | ||
| AK317713.1 | - | 4 | 494 | 135 | ||
| AY042830.1 | - | 4 | 466 | 135 | ||
| AY088335.1 | - | 4 | 472 | 135 | ||
More Information
Set Membership: |
|

RPS15A
Genomic Map