Assay Details
| Gene Symbol: RHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344637.1 | NP_001331800.1 | 2 | 1863 | 119 | |
| NM_001344638.1 | NP_001331801.1 | 2 | 1858 | 119 | ||
| NM_123938.4 | NP_199383.1 | 3 | 1657 | 119 | ||
| NM_203162.2 | NP_974891.1 | 2 | 1555 | 119 | ||
| GenBank mRNA | AK229058.1 | - | 3 | 1470 | 119 | |
More Information
Set Membership: |
|

RHA1
Genomic Map