Assay Details
| Gene Symbol: AT3G24508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035682.2 | NP_001030759.1 | 1 - 2 | 112 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G24508| Gene Symbol: AT3G24508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035682.2 | NP_001030759.1 | 1 - 2 | 112 | 79 | |
Set Membership: |
|