Assay Details
| Gene Symbol: SCRL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036539.2 | NP_001031616.1 | 1 - 2 | 83 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCRL21| Gene Symbol: SCRL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036539.2 | NP_001031616.1 | 1 - 2 | 83 | 110 | |
Set Membership: |
|