Assay Details
| Gene Symbol: ARPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036555.3 | NP_001031632.1 | 6 - 7 | 453 | 88 | |
| NM_001203797.1 | NP_001190726.1 | 7 - 8 | 486 | 88 | ||
| NM_001340903.1 | NP_001319932.1 | 7 - 8 | 671 | 88 | ||
More Information
Set Membership: |
|

ARPC4
Genomic Map