Assay Details
Gene Symbol: RIC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036663.2 | NP_001031740.1 | 1 - 2 | 721 | 81 | |
GenBank mRNA | DQ487576.1 | - | 1 - 2 | 71 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
