Assay Details
| Gene Symbol: AT5G19485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036831.4 | NP_001031908.1 | 8 - 9 | 924 | 121 | |
| GenBank mRNA | DQ487627.1 | - | 8 - 9 | 742 | 121 | |
More Information
Set Membership: |
|

AT5G19485
Genomic Map