Assay Details
| Gene Symbol: NOI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161310.1 | NP_001154782.1 | 3 - 4 | 478 | 101 | |
| NM_124967.3 | NP_200396.2 | 2 - 3 | 430 | 101 | ||
More Information
Set Membership: |
|

NOI
Genomic Map