Assay Details
| Gene Symbol: AT2G25500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128107.3 | NP_565598.3 | 1 | 157 | 140 | |
| GenBank mRNA | BT029245.1 | - | 1 | 157 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G25500| Gene Symbol: AT2G25500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128107.3 | NP_565598.3 | 1 | 157 | 140 | |
| GenBank mRNA | BT029245.1 | - | 1 | 157 | 140 | |
Set Membership: |
|