Assay Details
Gene Symbol: EDA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336584.1 | NP_001324434.1 | 3 - 4 | 626 | 116 | |
NM_129154.4 | NP_181139.2 | 3 - 4 | 602 | 116 | ||
GenBank mRNA | AK117562.1 | - | 3 - 4 | 581 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
