Assay Details
| Gene Symbol: PBC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036217.1 | NP_001031294.1 | 3 - 4 | 383 | 120 | |
| NM_106393.5 | NP_565156.1 | 4 - 5 | 516 | 120 | ||
| GenBank mRNA | AF043533.1 | - | 625 | 120 | ||
| AF385730.1 | - | 4 - 5 | 433 | 120 | ||
| AY081735.1 | - | 3 - 4 | 383 | 120 | ||
More Information
Set Membership: |
|

PBC2
Genomic Map