Assay Details
Gene Symbol: FUR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340534.1 | NP_001329797.1 | 4 - 5 | 538 | 153 | |
NM_116751.4 | NP_192421.1 | 5 - 6 | 669 | 153 | ||
GenBank mRNA | AF426400.1 | - | 4 - 5 | 445 | 153 | |
AK175753.1 | - | 5 - 6 | 526 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
