Assay Details
| Gene Symbol: FUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340534.1 | NP_001329797.1 | 5 - 6 | 639 | 152 | |
| NM_116751.4 | NP_192421.1 | 6 - 7 | 770 | 152 | ||
| GenBank mRNA | AF426400.1 | - | 5 - 6 | 546 | 152 | |
| AK175753.1 | - | 6 - 7 | 627 | 152 | ||
More Information
Set Membership: |
|

FUR1
Genomic Map