Assay Details
| Gene Symbol: BGLU31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343879.1 | NP_001331701.1 | 2 - 3 | 222 | 99 | |
| NM_122362.2 | NP_197842.1 | 2 - 3 | 221 | 99 | ||
| GenBank mRNA | DQ446980.1 | - | 2 - 3 | 221 | 99 | |
More Information
Set Membership: |
|

BGLU31
Genomic Map