Assay Details
Gene Symbol: BGLU31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343879.1 | NP_001331701.1 | 4 - 5 | 354 | 147 | |
NM_122362.2 | NP_197842.1 | 4 - 5 | 353 | 147 | ||
GenBank mRNA | DQ446980.1 | - | 4 - 5 | 353 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
