Assay Details
Gene Symbol: AT4G24370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118570.3 | NP_567700.1 | 1 | 452 | 54 | |
GenBank mRNA | AY063981.1 | - | 1 | 437 | 54 | |
AY086892.1 | - | 1 | 452 | 54 | ||
AY117195.1 | - | 1 | 423 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
