Assay Details
| Gene Symbol: TCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084312.1 | NP_001077781.1 | 1 | 754 | 147 | |
| NM_001160982.2 | NP_001154454.1 | 1 | 1154 | 147 | ||
| NM_001334285.1 | NP_001322103.1 | 1 | 1154 | 147 | ||
More Information
Set Membership: |
|

TCP1
Genomic Map