Assay Details
Gene Symbol: FHY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084549.2 | NP_001078018.1 | 1 - 2 | 151 | 95 | |
NM_001336669.1 | NP_001323653.1 | 1 - 2 | 151 | 95 | ||
GenBank mRNA | AF424739.1 | - | 1 - 2 | 54 | 95 | |
AK229717.1 | - | 1 - 2 | 151 | 95 | ||
HM029241.1 | - | 1 - 2 | 54 | 95 | ||
HM029242.1 | - | 1 - 2 | 54 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
