Assay Details
| Gene Symbol: FHY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084549.2 | NP_001078018.1 | 2 - 3 | 625 | 73 | |
| GenBank mRNA | AF424739.1 | - | 2 - 3 | 528 | 73 | |
| AK229717.1 | - | 2 - 3 | 625 | 73 | ||
| HM029242.1 | - | 2 - 3 | 528 | 73 | ||
More Information
Set Membership: |
|

FHY1
Genomic Map