Assay Details
| Gene Symbol: AT3G22142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084730.2 | NP_001078199.1 | 1 | 4151 | 101 | |
| Gene Symbol: AT3G22125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338565.1 | NP_001327427.1 | 1 | 559 | 101 | |
| NM_001338566.1 | NP_001327428.1 | 1 | 468 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001338565.1, NM_001338566.1 transcripts.
Set Membership: |
|

AT3G22142
Genomic Map