Assay Details
Gene Symbol: AT3G45248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084772.1 | NP_001078241.1 | 1 | 251 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AT3G45248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084772.1 | NP_001078241.1 | 1 | 251 | 112 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |