Assay Details
| Gene Symbol: DCI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085229.1 | NP_001078698.1 | 2 - 3 | 788 | 121 | |
| GenBank mRNA | AK316991.1 | - | 2 - 3 | 756 | 121 | |
More Information
Set Membership: |
|

DCI1
Genomic Map