Assay Details
| Gene Symbol: AT4G22513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084959.3 | NP_001078428.1 | 1 | 209 | 140 | |
| Gene Symbol: AT4G22510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341549.1 | NP_001320033.1 | 2 | 355 | 140 | |
| NM_001341550.1 | NP_001328263.1 | 2 | 459 | 140 | ||
More Information
Set Membership: |
|

AT4G22513
Genomic Map