Assay Details
| Gene Symbol: CNOT6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191523.2 | NP_001178452.1 | 5 - 6 | 619 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNOT6L| Gene Symbol: CNOT6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191523.2 | NP_001178452.1 | 5 - 6 | 619 | 67 | |
Set Membership: |
|