Assay Details
| Gene Symbol: MPLKIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075389.2 | NP_001068857.1 | 1 - 2 | 387 | 84 | |
| GenBank mRNA | BC120071.1 | - | 1 - 2 | 389 | 84 | |
More Information
Set Membership: |
|

MPLKIP
Genomic Map