Assay Details
Gene Symbol: PSMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034662.2 | NP_001029834.1 | 2 - 3 | 141 | 113 | |
GenBank mRNA | BC102206.1 | - | 2 - 3 | 142 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
