Assay Details
| Gene Symbol: EIF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037598.2 | NP_001032687.1 | 1 - 2 | 291 | 77 | |
| GenBank mRNA | BC109541.1 | - | 1 - 2 | 292 | 77 | |
More Information
Set Membership: |
|

EIF1B
Genomic Map