Assay Details
| Gene Symbol: DNAJC21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192218.3 | NP_001179147.1 | 1 - 2 | 255 | 57 | |
| GenBank mRNA | BC122750.1 | - | 1 - 2 | 262 | 57 | |
More Information
Set Membership: |
|

DNAJC21
Genomic Map