Assay Details
Gene Symbol: FKBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038112.3 | NP_001033201.1 | 3 - 4 | 336 | 118 | |
GenBank mRNA | BC109932.1 | - | 3 - 4 | 378 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
