Assay Details
| Gene Symbol: SLC22A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046502.2 | NP_001039967.1 | 8 - 9 | 1631 | 73 | |
| GenBank mRNA | BC105377.1 | - | 8 - 9 | 1633 | 73 | |
More Information
Set Membership: |
|

SLC22A5
Genomic Map