Assay Details
| Gene Symbol: GOSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038525.1 | NP_001033614.1 | 6 - 7 | 524 | 109 | |
| GenBank mRNA | BC109657.1 | - | 6 - 7 | 524 | 109 | |
More Information
Set Membership: |
|

GOSR1
Genomic Map