Assay Details
| Gene Symbol: PLPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080329.2 | NP_001073798.1 | 1 - 2 | 442 | 72 | |
| GenBank mRNA | BC109901.1 | - | 1 - 2 | 443 | 72 | |
More Information
Set Membership: |
|

PLPP1
Genomic Map