Assay Details
| Gene Symbol: RNF19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191331.1 | NP_001178260.1 | 4 - 5 | 1363 | 100 | |
| GenBank mRNA | BC151661.1 | - | 4 - 5 | 1342 | 100 | |
More Information
Set Membership: |
|

RNF19A
Genomic Map