Assay Details
| Gene Symbol: ZC3H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046394.2 | NP_001039859.1 | 3 - 4 | 390 | 79 | |
| GenBank mRNA | BC114825.1 | - | 3 - 4 | 391 | 79 | |
More Information
Set Membership: |
|

ZC3H15
Genomic Map