Assay Details
Gene Symbol: EGLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206046.2 | NP_001192975.2 | 2 - 3 | 1118 | 105 | |
GenBank mRNA | EE242828.1 | - | 2 - 3 | 565 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
