Assay Details
| Gene Symbol: PEA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075456.1 | NP_001068924.1 | 2 - 3 | 328 | 74 | |
| GenBank mRNA | BC119928.1 | - | 2 - 3 | 328 | 74 | |
More Information
Set Membership: |
|

PEA15
Genomic Map