Assay Details
| Gene Symbol: CNOT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034312.1 | NP_001029484.1 | 6 - 7 | 1004 | 93 | |
| GenBank mRNA | BC103000.1 | - | 6 - 7 | 1004 | 93 | |
More Information
Set Membership: |
|

CNOT7
Genomic Map