Assay Details
| Gene Symbol: ARPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034713.2 | NP_001029885.1 | 7 - 8 | 586 | 60 | |
| GenBank mRNA | BC105136.1 | - | 7 - 8 | 588 | 60 | |
More Information
Set Membership: |
|

ARPC2
Genomic Map