Assay Details
| Gene Symbol: TXNDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206732.1 | NP_001193661.1 | 1 - 2 | 299 | 51 | |
| GenBank mRNA | EH182135.1 | - | 1 - 2 | 299 | 51 | |
| EH205033.1 | - | 1 - 2 | 70 | 51 | ||
More Information
Set Membership: |
|

TXNDC5
Genomic Map