Assay Details
| Gene Symbol: UBE2G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082458.1 | NP_001075927.1 | 5 - 6 | 857 | 106 | |
| GenBank mRNA | BC133528.1 | - | 5 - 6 | 857 | 106 | |
More Information
Set Membership: |
|

UBE2G1
Genomic Map