Assay Details
| Gene Symbol: UBE2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038167.1 | NP_001033256.1 | 3 - 4 | 368 | 85 | |
| GenBank mRNA | BC111175.1 | - | 3 - 4 | 368 | 85 | |
More Information
Set Membership: |
|

UBE2D1
Genomic Map