Assay Details
| Gene Symbol: WASF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081511.1 | NP_001074980.1 | 2 - 3 | 283 | 84 | |
| GenBank mRNA | BC133284.1 | - | 2 - 3 | 283 | 84 | |
More Information
Set Membership: |
|

WASF2
Genomic Map