Assay Details
| Gene Symbol: RAB27B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174738.2 | NP_777163.1 | 4 - 5 | 664 | 110 | |
| GenBank mRNA | AY010899.1 | - | 4 - 5 | 664 | 110 | |
| BC122732.1 | - | 5 - 6 | 510 | 110 | ||
More Information
Set Membership: |
|

RAB27B
Genomic Map